ContactUsLinkComponent
C18G x 3µm particle size x 120Å pore size, 100mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-03M46P
C18G x 3µm particle size x 120Å pore size, 100mm length x 4.6mm ID, PEEK-coated ProteCol HPLC Column 1 * 1 items
Catalog # SGEA2C185-03M46P
Frequently Bought Together