ContactUsLinkComponent
Wiper 9x9Polyester Cellulose C30 is intended for use in critical cleanroom environments, such as microelectronics, semiconductor, food, pharmaceutical, cosmetics, and many other industries. C30 is strong, durable, highly absorbent and chemically pur 1 * 3.600 items
Catalog # EPTA7-C30-99P-10
Wiper 9x9Polyester Cellulose C30 is intended for use in critical cleanroom environments, such as microelectronics, semiconductor, food, pharmaceutical, cosmetics, and many other industries. C30 is strong, durable, highly absorbent and chemically pur 1 * 3.600 items
Catalog # EPTA7-C30-99P-10
Frequently Bought Together