ContactUsLinkComponent
Replacement lens for#611.02.00.00 goggle The sterilization in autoclave at 134°C for 7 min., or made by gamma or beta rays at 25kGy. item n° 04.20.60 1 * 1 items
Catalog # UNVT42060
Supplier:  UNIVET
Replacement lens for#611.02.00.00 goggle The sterilization in autoclave at 134°C for 7 min., or made by gamma or beta rays at 25kGy. item n° 04.20.60 1 * 1 items
Catalog # UNVT42060
Supplier:  UNIVET
Frequently Bought Together