Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Seed testing papers, Munktell

Seed testing papers, Munktell

292, 90 mm
N° de catalogue BINZ113311
Fournisseur:   Ahlstrom
Seed testing papers, Munktell
N° de catalogue BINZ113311
Fournisseur:  Ahlstrom
Seed testing papers, Munktell
Seed testing papers, Munktell

292, 90 mm

Prix
SFr. 8.60
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents

Seed Testing Papers are widely used by seed testing institutes and others for germination capacity testing and for health tests e.g. Jacobsen (Copenhagen tank), on top of paper and osmosis test.

Spécifications
  • Grade:
    292
  • Seq:
    11
  • Size:
    90 mm
  • Cat. No.:
    BINZ113311
  • Capillary rise:
    -
  • Ash content:
    -
  • Weight:
    87 g/m²