Accessories
Extended delivery jet assembly, 150 mm, for 25 ml dispenser
Katalognr. SOCO1.525.150
Anbieter: Socorex Isba
Direkt zu
Product Details & Documents
Technische Daten
- Description:Extended delivery jet assembly, 150 mm, for 25 ml dispenser
- Seq:19



