Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Accessories

Accessories

Delivery jet assembly, 120 mm, for 50 and 100 ml dispenser
Catalog # SOCO1.525.121
Supplier:   Socorex Isba
Accessories
Catalog # SOCO1.525.121
Supplier:  Socorex Isba
Accessories
Accessories

Delivery jet assembly, 120 mm, for 50 and 100 ml dispenser

Price
SFr. 75.10
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Delivery jet assembly, 120 mm, for 50 and 100 ml dispenser
  • Seq:
    16