Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

VWR® ATC, Handheld Refractometers

Refractometer for sugar, 0 − 55 °Bx, 1 °Bx
Catalog # 635-0634
Supplier:   VWR Collection
VWR® ATC, Handheld Refractometers
Catalog # 635-0634
Supplier:  VWR Collection
VWR® ATC, Handheld Refractometers
VWR® ATC, Handheld Refractometers

Refractometer for sugar, 0 − 55 °Bx, 1 °Bx

Price
SFr. 152.00
Log in for availability
1 * 1 ST
Product Details & Documents

Handheld refractometers with automatic temperature compensation (ATC) for consistent results.

  • ATC at 20 °C between 10 and 30 °C
  • Scales are easy to read and deliver exact results
  • Easy to use and clean
Specifications
  • Range:
    0 − 55 °Bx
  • Resolution:
    1 °Bx
  • Description:
    Refractometer for sugar
  • Seq:
    04
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Prism flap
    Prism flap
    635-0640
    Log in for availability
    1 * 1 ST - SFr. 29.20

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.