Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

VWR® ATC, Handheld Refractometers

Refractometer for sugar, 28 − 62 °Bx, 0,2 °Bx
Catalog # 635-0632
Supplier:   VWR Collection
VWR® ATC, Handheld Refractometers
Catalog # 635-0632
Supplier:  VWR Collection
VWR® ATC, Handheld Refractometers
VWR® ATC, Handheld Refractometers

Refractometer for sugar, 28 − 62 °Bx, 0,2 °Bx

Price
SFr. 152.00
Log in for availability
1 * 1 ST
Product Details & Documents

Handheld refractometers with automatic temperature compensation (ATC) for consistent results.

  • ATC at 20 °C between 10 and 30 °C
  • Scales are easy to read and deliver exact results
  • Easy to use and clean
Specifications
  • Range:
    28 − 62 °Bx
  • Resolution:
    0,2 °Bx
  • Description:
    Refractometer for sugar
  • Seq:
    05
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Prism flap
    Prism flap
    635-0640
    Log in for availability
    1 * 1 ST - SFr. 29.20