Jars with screw cap
150 ml
Codice catalogo 216-1570
Nome del fornitore: Anicrin
Vai a
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
- Twist cap for quick opening and closing
- Very good temperature resistance
- Very good chemical resistance
Attenzione: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifiche
- Capacity:150 ml
- Seq:01