Microincinerators, e-Loop
Product Details & Documents
Electrical sterilisation systems which are suitable for sterilising loops, needles and instruments. Ideal for use in laboratories where using gases or naked flames is not permitted. The e-Loop has a wear resistant quartz glass tube which is protected by a perforated stainless steel outer design. Choice of two units with different tube diameters, the larger is ideal for sterilising small instruments such as forceps and scissors. The heating system provides uniform heat. The opening eliminates the potential aerosol of pathogenic microorganisms.
- Loops and needles sterilised in 5 to 7 s
- Integral spray protector prevents hazardous gases, flames and infectious splatter harming user
- Sterilisation tube has self-locking angular adjustment, there are seven different positions
- Tray for three instruments
- Residual heat warning to prevent burns
: Optional accessories include a holder specifically for BioTool inoculation loop holders, a stainless steel tray which is positioned on the left hand side of the steriliser and stainless steel inoculation loop holders for loops with wire Ø 0,6 to 1 mm.
Specifications
- Model:710-1472
- Description:e-Loop Plant with 21 mm tube diameter
- Seq:02
- Cat. No.:710-1472
- W×D×H:175×160×173 mm
- Temperature range:900 - 950 °C
- Power supply:120 W
- Ø×L:21×161 mm
- Weight:1,6 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Stainless steel tray for positioning on left hand sideStainless steel tray for positioning on left hand side710-1575Log in for availability1 * 1 ST - SFr. 53.00
Product Information
- Catalog #AvailabilityPrice
- Holding device for BioTool loop holders
Product Information
- Catalog #AvailabilityPrice
- Quartz glass heating element for e-Loop 230 VQuartz glass heating element for e-Loop 230 V710-1573Log in for availability1 * 1 ST - SFr. 290.00
Product Information
- Catalog #AvailabilityPrice
- Spare ceramic annealing tube for e-Loop standard, all voltagesSpare ceramic annealing tube for e-Loop standard, all voltages710-1574Log in for availability1 * 1 ST - SFr. 77.00
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



