Orders LinkContactUsLinkComponent
Inserts
Inserts
E 484 1/1 insert, wire mesh for various utensils, 8 mm mesh spacing, G 7823, G 7824, G 7825, G 7826, PG 8528, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 150×470×480 mm
Catalog # MIEL5109190
Inserts
Catalog # MIEL5109190
Supplier:  
Inserts
Inserts
E 484 1/1 insert, wire mesh for various utensils, 8 mm mesh spacing, G 7823, G 7824, G 7825, G 7826, PG 8528, PG 8535/36, PG 8582, PG 8582 CD, PG 8592, 150×470×480 mm
Discontinued
Product Details & Documents
Specifications
  • Description:
    E 484 1/1 insert, wire mesh for various utensils, 8 mm mesh spacing
  • Seq:
    18
  • W×D×H:
    150×470×480 mm
  • For:
    G 7823, G 7824, G 7825, G 7826, PG 8528, PG 8535/36, PG 8582, PG 8582 CD, PG 8592