Biotix 250ul clear filter pipet tips for Beckman 96-well workstations. Racked, 10 racks of 96 Tips/Pack, 5 Packs/Case. AP96 Tips f or Beckman Biomek FX/NX, MultiMek 96, and Biomek 3000 workstations. Presterilized, RNase-, DNase-, and Endotoxin (py 1 * 4.800 items
Catalog # 732-2454
Supplier: BIOTIX
Frequently Bought Together