ContactUsLinkComponent
Biotix 1.5mL assorted colors microcentrifuge tube. 500 tubes/pack, 10 packs/case. Colors include blu/yel/grn/org/red. Max G force to 20,0000 RCF. Graduation marks for ID of liquid volume and syringe peirceable thin membrane caps. Autoclavable, RNa 1 * 500 items
Catalog # 525-0583
Supplier:  BIOTIX
undefined
Biotix 1.5mL assorted colors microcentrifuge tube. 500 tubes/pack, 10 packs/case. Colors include blu/yel/grn/org/red. Max G force to 20,0000 RCF. Graduation marks for ID of liquid volume and syringe peirceable thin membrane caps. Autoclavable, RNa 1 * 500 items
Catalog # 525-0583
Supplier:  BIOTIX
Frequently Bought Together