ContactUsLinkComponent
Biotix 1250 µL Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 10 Cards/Pack, 10 Pack/Case. Patented features: Fl exFit™ provides universal fit and superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, serial number 1 * 960 items
Catalog # 613-2822
Anbieter:  BIOTIX
Biotix 1250 µL Reload System Pipette Tips. Tip Eject Reload System 96 Tip/Card, 10 Cards/Pack, 10 Pack/Case. Patented features: Fl exFit™ provides universal fit and superior seal, X-Resin™ for low sample retention, Blade™ for accuracy, serial number 1 * 960 items
Catalog # 613-2822
Anbieter:  BIOTIX
Frequently Bought Together