ContactUsLinkComponent
Biotix 2.2mL 96 square well, V bottom, clear deep-well plate. 5 plates/pack, 10 packs/case. Alphanumeric grid and cut angle mark for tracking of samples. Polypropylene (PP), Presterilized, RNase-, DNase-, and Endotoxin (pyrogen)-free
Catalog # 732-2483
Supplier:  BIOTIX
Biotix 2.2mL 96 square well, V bottom, clear deep-well plate. 5 plates/pack, 10 packs/case. Alphanumeric grid and cut angle mark for tracking of samples. Polypropylene (PP), Presterilized, RNase-, DNase-, and Endotoxin (pyrogen)-free
Catalog # 732-2483
Supplier:  BIOTIX
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.