ContactUsLinkComponent
Biotix 10ul and 10XL LEER Pipette tip racks for Tip Eject Reload System. LEER racks come 10 Trays/Pack, 5 Packs/Case. These racks a re compatible with 10ul, 10XL, and 10ul Ultra Micro Tip Eject Pipette tips. 1 * 10 items
Katalognr. 613-2823
Supplier:  BIOTIX
Biotix 10ul and 10XL LEER Pipette tip racks for Tip Eject Reload System. LEER racks come 10 Trays/Pack, 5 Packs/Case. These racks a re compatible with 10ul, 10XL, and 10ul Ultra Micro Tip Eject Pipette tips. 1 * 10 items
Katalognr. 613-2823
Supplier:  BIOTIX
Frequently Bought Together