Basic Laboratory Chairs
The tried-and-tested all-rounder for use in laboratories is a great entry level laboratory chair, perfect for a range of workplace environments.
- With a sturdy steel 5 star base
- Available in two different frame colours: black or light grey
- Choice of three different mechanisms: contact backrest, contact backrest with seat tilt adjustment and synchronous mechanism with weight regulation
Basic Laboratory can be supplied with Stamskin synthetic leather or with fabric upholstery. Not only is Stamskin Top easy to clean, but it also has antibacterial, antifungal and antimicrobial properties.
Conform to: DIN 68 877, German GS „safety tested“ certificate and Quality Management System DIN EN ISO 9001
: Conform to: DIN 68 877, German GS 'safety tested' certificate and Quality Management System DIN EN ISO 9001
- Description:Basic laboratory 2 with synthetic leather upholstery, blue, backrest height: 530 mm, black frame
- Height:490 - 630 mm
- Seq:06
- Foot style:Castors
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



