Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Mesitylene for synthesis

Mesitylene for synthesis

165 kg
Catalog # 8.05890.6165
Flammable
Health
Irritant
Environment
Supplier:   Merck
Mesitylene for synthesis
Catalog # 8.05890.6165
Supplier:  Merck
CAS Number:  108-67-8
Mesitylene for synthesis

165 kg

CAS Number:  108-67-8
Price
Inquire For Price
Log in for availability
1 * 165 kg
Product Details & Documents
Synonyms:  1,3,5-Trimethylbenzene
  • CAS Number:108-67-8
  • Formula:C₉H₁₂
  • MDL Number:MFCD00008538
  • Molecular Weight:120.19 g/mol
  • Merck Index:13,05934
  • Boiling Pt:165 °C (1013 hPa)
  • Melting Pt:–45 °C
  • Density:0.8637 g/cm³ (20 °C)
  • Flash Pt:48 °C
  • Storage Temperature:Ambient
  • EINECS:203-604-4
  • UN:2325
  • ADR:3,III

Documents

Specifications

Specifications

  • Pk:
    165 kg

Specifications