LCTECH LITHIUM COLUMN REGENERANT 950ML 1 * 950 mL
Catalog # MSPPRG003
Supplier: MARKET SOURCE PART PROCESS
LCTECH LITHIUM COLUMN REGENERANT 950ML 1 * 950 mL
Catalog # MSPPRG003
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, EMSURE® for analysis, Supelco®
Supplier: Merck
Sodium sulphate, anhydrous 98.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Trione Ninhydrin Reagenz 1 * 1 items
Supplier: MARKET SOURCE PART PROCESS
LCTECH LITHIUM ELUENT PH 3.75 (4X950ML) 1 * 3.800 mL
Supplier: MARKET SOURCE PART PROCESS