HYDROCHLORIC ACID 0.1M 1 * 20 L
Catalog # REGEDB11-20
Supplier: REAGECON
Frequently Bought Together
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 60…80 °C, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
(±)-2-Butanol, EMPLURA®, Supelco®
Supplier: Merck
2-Propanol, extra dry 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific