ContactUsLinkComponent
Container to collect fluids. Capacity 500 L. Raw material : high density polyethylene. Colour : yellow. To collect leak or overflow of dangerous fluids. 1 * 1 items
Catalog # LAMA3297
Supplier:  LAMAPLAST
Container to collect fluids. Capacity 500 L. Raw material : high density polyethylene. Colour : yellow. To collect leak or overflow of dangerous fluids. 1 * 1 items
Catalog # LAMA3297
Supplier:  LAMAPLAST
Frequently Bought Together