ContactUsLinkComponent
2mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 2mg/L (RTOCS03) TOC C as Sucrose and 2mg/L (RTICN04) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120017
Supplier:  REAGECON
2mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 2mg/L (RTOCS03) TOC C as Sucrose and 2mg/L (RTICN04) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120017
Supplier:  REAGECON
Frequently Bought Together