ContactUsLinkComponent
0.5mg/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 0.5mg/L (RTOCS01) TOC C as Sucrose an d 0.5mg/L (RTICN01) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120008
Supplier:  REAGECON
0.5mg/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 0.5mg/L (RTOCS01) TOC C as Sucrose an d 0.5mg/L (RTICN01) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120008
Supplier:  REAGECON
Frequently Bought Together