ContactUsLinkComponent
CYP2D6.2 BACTOSOMES CYP2D6.2 BACTOSOMES EASYCYP CYP2D6X2R HUMAN CYP2D6*2R EASYCYP BACTOSOMES HUMAN CYP2D6.2 AND HUMAN CYP-REDUCTASE COEXPRESSED IN ESCHERICHIA COLI P450 ACTIVITY DATA - 24 PMOL/MIN/PMOL CY
Catalog # CYPEEZ027
Supplier:  CYPEX
CYP2D6.2 BACTOSOMES CYP2D6.2 BACTOSOMES EASYCYP CYP2D6X2R HUMAN CYP2D6*2R EASYCYP BACTOSOMES HUMAN CYP2D6.2 AND HUMAN CYP-REDUCTASE COEXPRESSED IN ESCHERICHIA COLI P450 ACTIVITY DATA - 24 PMOL/MIN/PMOL CY
Catalog # CYPEEZ027
Supplier:  CYPEX
Frequently Bought Together