ContactUsLinkComponent
Power supply economical, UK, for ECO27 spectral lamp with ECO27 screw sockets, adapted ventilation, 2P+E power outlet with fuse, no risk of burning, economical, can be used on the lamp base or on optical bench. 1 * 1 items
Catalog # DIDAPOF010060_UK
Supplier:  DIDALAB
Power supply economical, UK, for ECO27 spectral lamp with ECO27 screw sockets, adapted ventilation, 2P+E power outlet with fuse, no risk of burning, economical, can be used on the lamp base or on optical bench. 1 * 1 items
Catalog # DIDAPOF010060_UK
Supplier:  DIDALAB
Frequently Bought Together