ContactUsLinkComponent
Viewfinder for the accurate measurement of focal lengths, high precision crank and pinion, objective 4x, supplied with two 10x eyepi eces (1/10 mm micrometric eyepieces with a cross-shaped reticle). 1 * 1 items
Catalog # DIDAPOD069440
Supplier:  DIDALAB
Viewfinder for the accurate measurement of focal lengths, high precision crank and pinion, objective 4x, supplied with two 10x eyepi eces (1/10 mm micrometric eyepieces with a cross-shaped reticle). 1 * 1 items
Catalog # DIDAPOD069440
Supplier:  DIDALAB
Frequently Bought Together