ContactUsLinkComponent
Slit adjustable - from 0 to 8mm. effective length of 42mm. intended for experiments that do not require the knowledge of the opening , this slit can be mounted horizontally or vertically. 1 * 1 items
Catalog # DIDAPOM051540
Supplier:  DIDALAB
Slit adjustable - from 0 to 8mm. effective length of 42mm. intended for experiments that do not require the knowledge of the opening , this slit can be mounted horizontally or vertically. 1 * 1 items
Catalog # DIDAPOM051540
Supplier:  DIDALAB
Frequently Bought Together