ContactUsLinkComponent
Leakage current tester, DCI/ACI : 25ma~50µa, equipment under test, suitable for medical electrical and general of leakage current me asurement, touch panel with color display, 9 different measurements to simulate the resistance of h 1 * 1 items
Catalog # DIDAPMD500020
Supplier:  DIDALAB
Leakage current tester, DCI/ACI : 25ma~50µa, equipment under test, suitable for medical electrical and general of leakage current me asurement, touch panel with color display, 9 different measurements to simulate the resistance of h 1 * 1 items
Catalog # DIDAPMD500020
Supplier:  DIDALAB
Frequently Bought Together