ContactUsLinkComponent
Mercury spectral lamp, power supply + low pressure, 6W, adapted ventilation, 2P+E power outlet with fuse, no risk of burning, econom ical, can be used on the lamp base or on optical bench, UK. 1 * 1 items
Catalog # DIDAPOD010050_UK
Supplier:  DIDALAB
Mercury spectral lamp, power supply + low pressure, 6W, adapted ventilation, 2P+E power outlet with fuse, no risk of burning, econom ical, can be used on the lamp base or on optical bench, UK. 1 * 1 items
Catalog # DIDAPOD010050_UK
Supplier:  DIDALAB
Frequently Bought Together