Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Optical components for optical bench

Neutral density filter, d=0,9, set in an M49 ring
  DIDAPOD010021
 :   DIDALAB
Optical components for optical bench
  DIDAPOD010021
 :  DIDALAB
Optical components for optical bench
Optical components for optical bench

Neutral density filter, d=0,9, set in an M49 ring

 
SFr. 26.60
   
1 * 1 ST
 
 
  • Description:
    Neutral density filter, d=0,9, set in an M49 ring
Accessories

 

  •  
    Description
     
     
  • DIDAPOD010095
    Permanent frame, Ø 40 mm
       
    1 * 1 ST - SFr. 40.80

 

  •  
    Description
     
     
  • DIDAPOD010110
    Simple lens holder, Ø 40 mm
       
    1 * 1 ST - SFr. 20.00

 

  •  
    Description
     
     
  • DIDAPOD010080
    Lens holder for fragile components, Ø 80 mm
       
    1 * 1 ST - SFr. 71.50

 

  •  
    Description
     
     
  • DIDAPOD010090
    Lens holder, Ø 40/42 mm
       
    1 * 1 ST - SFr. 37.50

 

  •  
    Description
     
     
  • DIDAPOD060130
    Lens holder, Ø 80 mm
       
    1 * 1 ST - SFr. 57.40

 

  •  
    Description
     
     
  • DIDAPOD002192
    Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
       
    1 * 1 ST - SFr. 120.00