ContactUsLinkComponent
RLC meter, 20,000/2,000 counts dual display, test frequency 100/120hz/1khz/10khz, 0.2% basic accuracy, parallel /series testing mode , 2-wire or 5-wire measurement available, data hold, max, min and zero mode supported, auto cklit, UK. 1 * 1 items
Catalog # DIDAPMD400040_UK
Supplier:  DIDALAB
RLC meter, 20,000/2,000 counts dual display, test frequency 100/120hz/1khz/10khz, 0.2% basic accuracy, parallel /series testing mode , 2-wire or 5-wire measurement available, data hold, max, min and zero mode supported, auto cklit, UK. 1 * 1 items
Catalog # DIDAPMD400040_UK
Supplier:  DIDALAB
Frequently Bought Together