ContactUsLinkComponent
Michelson interferometer, motorized, diameter 80mm beamsplitters, with anti-relfective coating, diameter 40mm mirrors, planeity of t he beamsplitters and mirrors λ/20, we put the orientation and lead systems of the compensator beamsp 1 * 1 items
Codice catalogo DIDAPOD013493
Nome del fornitore:  DIDALAB
Michelson interferometer, motorized, diameter 80mm beamsplitters, with anti-relfective coating, diameter 40mm mirrors, planeity of t he beamsplitters and mirrors λ/20, we put the orientation and lead systems of the compensator beamsp 1 * 1 items
Codice catalogo DIDAPOD013493
Nome del fornitore:  DIDALAB
Frequently Bought Together