ContactUsLinkComponent
Lighting kit for the michelson interferometer, consist of 1 50cm optical bench, 3 staples, 1 lamp with variator, 2 lens holders (for diameter 80mm lenses) , 1 diameter 80mm lens, f +150mm, 1 diameter 80mm lens, f +250mm, 1 diaph er, UK. 1 * 1 items
Catalog # DIDAPOD013492_UK
Supplier:  DIDALAB
Lighting kit for the michelson interferometer, consist of 1 50cm optical bench, 3 staples, 1 lamp with variator, 2 lens holders (for diameter 80mm lenses) , 1 diameter 80mm lens, f +150mm, 1 diameter 80mm lens, f +250mm, 1 diaph er, UK. 1 * 1 items
Catalog # DIDAPOD013492_UK
Supplier:  DIDALAB
Frequently Bought Together