ContactUsLinkComponent
Michelson interferometer, diameter 80mm beamsplitters, with anti-relfective coating, diameter 40mm mirrors, planeity of the beamspli tters and mirrors λ/20, we put the orientation and lead systems of the compensator beamsplitter. 1 * 1 items
Catalog # DIDAPOD013490
Supplier:  DIDALAB
Michelson interferometer, diameter 80mm beamsplitters, with anti-relfective coating, diameter 40mm mirrors, planeity of the beamspli tters and mirrors λ/20, we put the orientation and lead systems of the compensator beamsplitter. 1 * 1 items
Catalog # DIDAPOD013490
Supplier:  DIDALAB
Frequently Bought Together