Order Entry
Switzerland
Orders LinkContactUsLinkComponent
High visibility rain jacket, Essential, class 3

High visibility rain jacket, Essential, class 3

Hi-vis orange, M
Catalog # KWIN101012230M
Supplier:   FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
Catalog # KWIN101012230M
Supplier:  FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
High visibility rain jacket, Essential, class 3

Hi-vis orange, M

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Basic rain jacket made of polyurethane-coated nylon jersey with 'stretch' function.

  • Detachable drawstring hood
  • Buttons and zip
  • Adjustable cuffs

Weight: 170 g/m²

Certifications: EN 471 (3-2), EN 343 (3-2)

Documents

Specifications
  • Clothing Size:
    M
  • Colour:
    Hi-vis orange
  • Seq:
    12
  • Cat. No.:
    KWIN101012230M
  • Sleeve or leg length:
    Long
  • Risk area:
    Rain (waterproof)
  • Model:
    Class 3
  • Garment application:
    General Purpose
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Fabric:
    Polyurethane-coated nylon
  • Weight:
    170 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.