Order Entry
Switzerland
Orders LinkContactUsLinkComponent
High visibility rain jacket, Essential, class 3 | Avantor

High visibility rain jacket, Essential, class 3

Hi-vis yellow, XS
Katalognr. KWIN1010121301XS
Anbieter:   FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
Katalognr. KWIN1010121301XS
Anbieter:  FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
High visibility rain jacket, Essential, class 3

Hi-vis yellow, XS

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

Basic rain jacket made of polyurethane-coated nylon jersey with 'stretch' function.

  • Detachable drawstring hood
  • Buttons and zip
  • Adjustable cuffs

Weight: 170 g/m²

Zertifizierung: EN 471 (3-2), EN 343 (3-2)

Dokumente

Technische Daten
  • Clothing Size:
    XS
  • Colour:
    Hi-vis yellow
  • Seq:
    02
  • Cat. No.:
    KWIN1010121301XS
  • Sleeve or leg length:
    Long
  • Risk area:
    Rain (waterproof)
  • Model:
    Class 3
  • Garment application:
    General Purpose
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Fabric:
    Polyurethane-coated nylon
  • Weight:
    170 g/m²
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.