Order Entry
Switzerland
Orders LinkContactUsLinkComponent
High visibility rain jacket, Essential, class 3

High visibility rain jacket, Essential, class 3

Hi-vis orange, XXL
Catalog # KWIN1010122302XL
Supplier:   FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
Catalog # KWIN1010122302XL
Supplier:  FRISTADS KANSAS
High visibility rain jacket, Essential, class 3
High visibility rain jacket, Essential, class 3

Hi-vis orange, XXL

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Basic rain jacket made of polyurethane-coated nylon jersey with 'stretch' function.

  • Detachable drawstring hood
  • Buttons and zip
  • Adjustable cuffs

Weight: 170 g/m²

Certifications: EN 471 (3-2), EN 343 (3-2)

Documents

Specifications
  • Clothing Size:
    XXL
  • Colour:
    Hi-vis orange
  • Seq:
    15
  • Cat. No.:
    KWIN1010122302XL
  • Sleeve or leg length:
    Long
  • Risk area:
    Rain (waterproof)
  • Model:
    Class 3
  • Garment application:
    General Purpose
  • Gender:
    Unisex
  • Closure type:
    Zip closure
  • Fabric:
    Polyurethane-coated nylon
  • Weight:
    170 g/m²