KCL ELECTROLYTE SOLUTION CPY4 3.0 MOL 1 * 250 mL
Catalog # ENDRCPY4-5
Supplier: Endress+Hauser
Frequently Bought Together
Stainless Steel Buckets
Supplier: BOCHEM
VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
VWR®, Reagents in tablet form for photometric determination of chlorine
Supplier: VWR Chemicals
Screw Caps for Wide Neck Bottles
Supplier: RIXIUS
Cryotube rack
Supplier: Brand
Qualitative filter papers, standard grades, grade 1 and 1V, Whatman™
Supplier: Whatman products (Cytiva)
Macro Cells with PTFE Stopper for Absorption Measurements
Supplier: HELLMA
Centrefeed rolls and wipes, Tork
Supplier: Essity
VWR®, Permanent Marker Pens
Supplier: VWR Collection
pH electrodes, SenTix®
Supplier: WTW
Acetic acid 99 - 100% 99-100%, GPR RECTAPUR®
Supplier: VWR Chemicals
Cartridge for bunsen burner, LABOGAZ
Supplier: GLASWARENFABRIK KARL HECHT
VWR®, Bottles, HDPE
Supplier: VWR Collection
Acetaldehyde 99.5%, extra pure
Supplier: Thermo Fisher Scientific
Accessory for storing electrodes correctly 1 * 1 items
Supplier: HAMILTON BONADUZ
Sodium tetraethylborate 97%, pure
Supplier: Thermo Fisher Scientific
Buffer solution pH 4.006 @25?C NIST and DIN 16266, 6x 250ml, Mettler 1 * 6 items
Supplier: Mettler - Toledo
Bottles, narrow or wide neck, square or rectangular, with thread
Supplier: RIXIUS