4-Vinyl-1,3-dioxolan-2-one
50 g
Supplier: Thermo Fisher Scientific
4-Vinyl-1,3-dioxolan-2-one
Catalog # H60822.18
Supplier: Thermo Fisher Scientific
CAS Number: 4427-96-7
Jump to:
Product Details & Documents
- CAS Number:4427-96-7
- Formula:C₅H₆O₃
- MDL Number:MFCD00143315
- Molecular Weight:114.10 g/mol
- Boiling Pt:237 °C
- Density:1.188
- Flash Pt:96 °C (205 °F)
- Flash Pt:96 °C(205 °F)
- Storage Temperature:Ambient
- UN:2810
- ADR:6.1,III
- ADR:"6.1",III
Documents
Specifications
Specifications
- Pk:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



