Mops, LoopMop™
LoopMop™, irradiated
Katalognr. MICRMZLM5-12IR
Anbieter: MICRONOVA
Direkt zu
Product Details & Documents
Knitted polyester mop for applying disinfectant in pharmaceutical, aseptic processing and medical device applications. Absorbent enough to clean to a dry finish but will leave enough moisture for disinfectants that require an extended drying time. Mop heads can be used directly with the fibreglass/polypropylene handle or attached to the universal handle via the quick disconnect adapter.
- Can be laundered or autoclaved
- Low linting
- No raw edges
Informazioni per l'ordine: All components must be ordered separately.
Technische Daten
- Description:LoopMop™, irradiated
- Cat. No.:MICRMZLM5-12IR
- Environmentally Preferable:Waste Reducing
Accessories
Produktinformationen
- Katalognr.DescriptionLengthMaterialVerfügbarkeitPreis
- 129-0551Stainless steel quick disconnect extendable handle965 - 1829 mmStainless steel 304Einloggen für Verfügbarkeit1 * 1 ST - SFr. 449.00
Produktinformationen
- Katalognr.DescriptionLengthMaterialVerfügbarkeitPreis
- 129-0062Stainless steel quick disconnect handle155 cmFibreglassEinloggen für Verfügbarkeit1 * 1 ST - SFr. 207.00
Produktinformationen
- Katalognr.DescriptionLengthMaterialVerfügbarkeitPreis
- 129-0176Stainless steel quick disconnect handle1295 cmStainless steel 304Einloggen für Verfügbarkeit1 * 1 ST - SFr. 200.00
Produktinformationen
- Katalognr.DescriptionVerfügbarkeitPreis
- 129-0170MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handlesEinloggen für Verfügbarkeit1 * 1 ST - SFr. 197.00