Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Bucket Liners

Bucket Liners

Disposable bucket liners, double bagged, irradiated, LDPE/nylon, 457,2×457,2 mm
Catalog # MICRBKL1-1818IR
   Waste Reducing
Supplier:   MICRONOVA
Bucket Liners
Catalog # MICRBKL1-1818IR
Supplier:  MICRONOVA
   Waste Reducing
Bucket Liners
Bucket Liners

Disposable bucket liners, double bagged, irradiated, LDPE/nylon, 457,2×457,2 mm

Price
SFr. 645.00
Log in for availability
1 * 50 ST
Product Details & Documents

The unique LDPE/nylon laminate is a good barrier material for transporting raw materials or to line buckets containing disinfectants or other cleaning solutions.

  • Laminate is 75% LDPE, 25% nylon
  • Excellent for disinfectants and cleaning solutions
  • Ideal for product parts transport

Designed for cleanrooms and controlled environments. Available as sterile and non-irradiated.

Packaging: Individually double packed, packed in a case of 24/50/200.

Specifications
  • Description:
    Disposable bucket liners, double bagged, irradiated
  • L×W:
    457,2×457,2 mm
  • Material:
    LDPE/nylon
  • Seq:
    01
  • Cat. No.:
    MICRBKL1-1818IR
  • Environmentally Preferable:
    Waste Reducing
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.