ContactUsLinkComponent
Gadolinium Gd - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM321.2NP.L1
Supplier:  CPACHEM
Gadolinium Gd - 100 mg/l Standard for ICP/MS in HNO3 1 * 100 mL
Catalog # ACSDM321.2NP.L1
Supplier:  CPACHEM
Frequently Bought Together