Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Multi-element quality control standards for ICP

Multi-element quality control standards for 33 elements, 5% HNO₃, 500 mL
Catalog # ACSDM8A96.1.5N.L5
Supplier:   CPACHEM
Multi-element quality control standards for ICP
Catalog # ACSDM8A96.1.5N.L5
Supplier:  CPACHEM
CAS Number:  
Multi-element quality control standards for ICP
Multi-element quality control standards for ICP

Multi-element quality control standards for 33 elements, 5% HNO₃, 500 mL

CAS Number:  
Price
SFr. 407.00
Log in for availability
1 * 500 mL
Product Details & Documents

ICP QC standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn. 1 mg/l for 33 elememts.

 

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    500 mL
  • Description:
    Multi-element quality control standards for 33 elements

Specifications