Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Multi-element quality control standards for ICP

Multi-element quality control standards for ICP

Multi-element quality control standards for 33 elements, 5% HNO₃, 100 mL
Catalog # ACSDM8A96.1.5N.L1
Supplier:   CPACHEM
Multi-element quality control standards for ICP
Catalog # ACSDM8A96.1.5N.L1
Supplier:  CPACHEM
CAS Number:  
Multi-element quality control standards for ICP
Multi-element quality control standards for ICP

Multi-element quality control standards for 33 elements, 5% HNO₃, 100 mL

CAS Number:  
Price
SFr. 168.00
Log in for availability
1 * 100 mL
Product Details & Documents

ICP QC standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn. 1 mg/l for 33 elememts.

Documents

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element quality control standards for 33 elements

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.