[EN]LEAD WIRE W/BANANA PLUG 1 * 2 ST
Catalog # PARRA297E
Supplier: PARR
Frequently Bought Together
Manschette für Röhrchen 4 - 6 mm ID 1 * 1 ST
Supplier: INTEGRA
KIT RAPID DISPENSING FOR MEDIACLAVE 30 1 * 1 items
Supplier: INTEGRA
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Colorimetrics tubes for gas measurements
Supplier: GASTEC
VWR®, Eluent buffers for amino acid analyzer
Supplier: VWR Chemicals
Gas Chromatograph with FID (GC-FID)
Supplier: LUCIDITY
VWR®, Gravimetric Dilutor, 3 kg
Supplier: VWR Collection
Carousel tissue processors
Supplier: Slee
Folding magnifier, metal
Supplier: ESCHENBACH OPTIK
Glass refill for dish-shaped dewar flasks SCH-CAL
Supplier: KGW
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Safety air inlet valves
Supplier: VICI JOUR RESEARCH
MEASURING CELL | M-VROC II C20-CHIP 1 * 1 ST
Supplier: MARKET SOURCE PART PROCESS
Oils for rotational viscometers, ROTAVISC
Supplier: IKA
[EN]VERTICAL HOLDER FOR TABLE OR TRIPOD 1 * 1 ST
Supplier: CUSTOM MADE CHEMICALS
Accessories for X-act® 5000
Supplier: Dräger