ContactUsLinkComponent
SNRPE (Human) - 3 unique 27mer siRNA duplexes - 2 nmol each (Locus ID 6635) Included - SR30004, Trilencer-27 Universal Scrambled Negative Control siRNA Duplex - 2 nmol Included - SR30005, RNAse free siRNA Duplex Resuspension Buffer - 2 ml 1 * 1 KIT
Catalog # ORIGSR304508
Supplier:  OriGene
SNRPE (Human) - 3 unique 27mer siRNA duplexes - 2 nmol each (Locus ID 6635) Included - SR30004, Trilencer-27 Universal Scrambled Negative Control siRNA Duplex - 2 nmol Included - SR30005, RNAse free siRNA Duplex Resuspension Buffer - 2 ml 1 * 1 KIT
Catalog # ORIGSR304508
Supplier:  OriGene
Frequently Bought Together