Single channel pipettes, mechanical, variable volume, VITLAB®
1000 - 10000 µl, ±0,6 %, ≤0,2 %, 10000 µl
Catalog # VITL1641012
Supplier: VITLAB
Single channel pipettes, mechanical, variable volume, VITLAB®
Catalog # VITL1641012
Supplier: VITLAB
Jump to:
Product Details & Documents
These piston-operated manual pipettes are ideal for demanding laboratory applications.
- Central, large pipetting button for easy, uniform piston movement
- Large, ergonomic eject button for rapid replacement of the tips
- Simple volume adjustment for right- and left-handers
- Precise, 4-position volume indicator, with integrated zoom function for optimal readability
- Colour-coded for easy selection of the right pipette tip
- Ergonomic finger loop for convenient handling
- Integrated calibration function for easy calibration without the need for tools
- Corrosion-resistant pistons and ejectors for long product life
- Completely autoclavable at 121 °C
DE-M marked
Delivery information: Supplied with silicone oil resp. grease (Size 5 and 10 ml), sample bag with pipette tips, quality certificate, and instruction manual
Specifications
- Volume:1000 - 10000 µl
- Accuracy:±0,6 %
- Imprecision:≤0,2 %
- Recommended tips:10000 µl
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..



