Recombinant Human sCD27 Ligand 1 * 10 µG
Catalog # PEPR310-30-10UG
Anbieter: PeproTech, Inc.
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Anbieter: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Anbieter: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Anbieter: HEATHROW
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Anbieter: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Anbieter: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Anbieter: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Anbieter: VWR Chemicals
VWR®, Nitrile Gloves
Anbieter: VWR Collection
VWR® Facial Tissues
Anbieter: VWR Collection
pH indicator strips, MQuant®, Supelco®
Anbieter: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Anbieter: VWR Chemicals