ContactUsLinkComponent
Human -NGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinan t human -NGF in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reag 1 * 200 Tests
Catalog # PEPR900-M60
Supplier:  PeproTech, Inc.
Human -NGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinan t human -NGF in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, reag 1 * 200 Tests
Catalog # PEPR900-M60
Supplier:  PeproTech, Inc.
Frequently Bought Together