ContactUsLinkComponent
Murine IL-17 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombina nt murine IL-17 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, re 1 * 200 Tests
Katalognr. PEPR900-M392
Supplier:  PeproTech, Inc.
Murine IL-17 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombina nt murine IL-17 in a sandwich ELISA format. Using the ELISA protocol described below, the recommended microplates, re 1 * 200 Tests
Katalognr. PEPR900-M392
Supplier:  PeproTech, Inc.
Frequently Bought Together